trans-2-(3-Fluorophenyl)vinylboronic acid 95%

Lieferant: Apollo Scientific

PC901430-1G PC901430-250MG
APOSPC901430-1GEA 0 EUR
APOSPC901430-1G APOSPC901430-250MG
trans-2-(3-Fluorophenyl)vinylboronic acid 95%
trans-2-(3-Fluorophenyl)vinylboronic acid
Formel: C₈H₈BFO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06008297
CAS-Nummer: 849062-22-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR