1-Methyl-3-trifluoromethylpyrazole-4-boronic acid 95%

Lieferant: Apollo Scientific

Synonyme: (1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-yl)boronic acid

PC901442-1G
APOSPC901442-1GEA 116 EUR
APOSPC901442-1G
1-Methyl-3-trifluoromethylpyrazole-4-boronic acid 95%
1-Methyl-3-trifluoromethylpyrazole-4-boronic acid
Formel: C₅H₆BF₃N₂O₂
Molecular Weight: 193.92 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13619875
CAS-Nummer: 1138450-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR