N-Ethyl-4-fluorbenzamid 98%

Lieferant: Apollo Scientific

PC901487-100G PC901487-25G PC901487-5G
APOSPC901487-100GEA 899 EUR
APOSPC901487-100G APOSPC901487-25G APOSPC901487-5G
N-Ethyl-4-fluorbenzamid 98%
N-Ethyl-4-fluorbenzamid
Formel: C₉H₁₀FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD01214153
CAS-Nummer: 772-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR