N-Ethyl-3-fluorbenzamid 98%

Lieferant: Apollo Scientific

PC901491-100G PC901491-25G PC901491-5G
APOSPC901491-100GEA 632 EUR
APOSPC901491-100G APOSPC901491-25G APOSPC901491-5G
N-Ethyl-3-fluorbenzamid 98%
N-Ethyl-3-fluorbenzamid
Formel: C₉H₁₀FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD01338769
CAS-Nummer: 150079-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR