4-Fluoro-2,3-dimethylbenzoic acid 95%
Lieferant: Apollo Scientific
PC901515-1G
APOSPC901515-1GEA
632
EUR
APOSPC901515-1G
4-Fluoro-2,3-dimethylbenzoic acid 95%
4-Fluoro-2,3-dimethylbenzoic acid
Formel:
C₉H₉FO₂ Molecular Weight: 168,165 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23712108 CAS-Nummer: 1104380-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...