Methyl 3-amino-5-(trifluoromethyl)benzoate 97%

Lieferant: Apollo Scientific

PC901522-25G
APOSPC901522-25GEA 266 EUR
APOSPC901522-25G
Methyl 3-amino-5-(trifluoromethyl)benzoate 97%
Methyl 3-amino-5-(trifluoromethyl)benzoate
Formel: C₉H₈F₃NO₂
Molecular Weight: 219.161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00625827
CAS-Nummer: 22235-25-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR