4-Ethoxy-2-fluorobenzaldehyde 98%

Lieferant: Apollo Scientific

PC901566-1G
APOSPC901566-1GEA 270 EUR
APOSPC901566-1G
4-Ethoxy-2-fluorobenzaldehyde 98%
4-Ethoxy-2-fluorobenzaldehyde
Formel: C₉H₉FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06247347
CAS-Nummer: 532965-74-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR