1-(2-Fluoro-5-methoxyphenyl)ethanone 96%

Lieferant: Apollo Scientific

PC901592-1G
APOSPC901592-1GEA 369 EUR
APOSPC901592-1G
1-(2-Fluoro-5-methoxyphenyl)ethanone 96%
1-(2-Fluoro-5-methoxyphenyl)ethanone
Formel: C₉H₉FO₂
Molecular Weight: 168,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01074363
CAS-Nummer: 80309-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR