Methyl 5-cyano-2-fluorobenzoate 95%

Lieferant: Apollo Scientific

Synonyme: 5-Cyano-2-fluorobenzoic acid methyl ester

PC901594-10G
APOSPC901594-10GEA 960 EUR
APOSPC901594-10G
Methyl 5-cyano-2-fluorobenzoate 95%
Methyl 5-cyano-2-fluorobenzoate
Formel: C₉H₆FNO₂
Molecular Weight: 179,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11042790
CAS-Nummer: 337362-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR