3,5-Difluoro-4-hydroxybenzoic acid 97%

Lieferant: Apollo Scientific

PC901597-1G
APOSPC901597-1GEA 1350 EUR
APOSPC901597-1G
3,5-Difluoro-4-hydroxybenzoic acid 97%
3,5-Difluoro-4-hydroxybenzoic acid
Formel: C₇H₄F₂O₃
Molecular Weight: 174.102 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12406861
CAS-Nummer: 74799-63-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR