Methyl 2,6-difluoro-3-hydroxybenzoate 98%

Lieferant: Apollo Scientific

PC901603-1G
APOSPC901603-1GEA 806 EUR
APOSPC901603-1G
Methyl 2,6-difluoro-3-hydroxybenzoate 98%
Methyl 2,6-difluoro-3-hydroxybenzoate
Formel: C₈H₆F₂O₃
Molecular Weight: 188.128 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14698033
CAS-Nummer: 1214332-41-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR