2,4-difluoro-3-hydroxybenzoic acid 95%

Lieferant: Apollo Scientific

PC901612-1G PC901612-250MG
APOSPC901612-1GEA 180 EUR
APOSPC901612-1G APOSPC901612-250MG
2,4-difluoro-3-hydroxybenzoic acid 95%
2,4-difluoro-3-hydroxybenzoic acid
Formel: C₇H₄F₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD06797839
CAS-Nummer: 91659-08-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR