2,4-difluoro-3-hydroxybenzoic acid 95%
Lieferant: Apollo Scientific
PC901612-1G
PC901612-250MG
APOSPC901612-1GEA
180
EUR
APOSPC901612-1G
APOSPC901612-250MG
2,4-difluoro-3-hydroxybenzoic acid 95%
2,4-difluoro-3-hydroxybenzoic acid
Formel:
C₇H₄F₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797839 CAS-Nummer: 91659-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...