2-Ethoxy-5-fluorobenzonitrile 96%

Lieferant: Apollo Scientific

PC901613-1G
APOSPC901613-1GEA 0 EUR
APOSPC901613-1G
2-Ethoxy-5-fluorobenzonitrile 96%
2-Ethoxy-5-fluorobenzonitrile
Formel: C₉H₈FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD15201675
CAS-Nummer: 1158201-03-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR