Methyl 3-(3-fluorophenyl)propanoate 98%
Lieferant: Apollo Scientific
PC901617-1G
PC901617-5G
APOSPC901617-1GEA
155
EUR
APOSPC901617-1G
APOSPC901617-5G
Methyl 3-(3-fluorophenyl)propanoate 98%
Methyl 3-(3-fluorophenyl)propanoate
Formel:
C₁₀H₁₁FO₂ Molecular Weight: 182.192 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12031749 CAS-Nummer: 425704-52-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...