Methyl 3-(3-fluorophenyl)propanoate 98%

Lieferant: Apollo Scientific

PC901617-1G PC901617-5G
APOSPC901617-1GEA 155 EUR
APOSPC901617-1G APOSPC901617-5G
Methyl 3-(3-fluorophenyl)propanoate 98%
Methyl 3-(3-fluorophenyl)propanoate
Formel: C₁₀H₁₁FO₂
Molecular Weight: 182.192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12031749
CAS-Nummer: 425704-52-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR