6-Fluoro-4,4-dimethyl-2,3-dihydro-1H-quinoline 95%
Lieferant: Apollo Scientific
PC901618-1G
APOSPC901618-1GEA
574
EUR
APOSPC901618-1G
6-Fluoro-4,4-dimethyl-2,3-dihydro-1H-quinoline 95%
6-Fluoro-4,4-dimethyl-2,3-dihydro-1H-quinoline
Formel:
C₁₁H₁₄FN Molecular Weight: 179,234 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12913850 CAS-Nummer: 345264-92-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...