3,3,3-Trifluoropropanamide 95%

Lieferant: Apollo Scientific

PC901625-1G
APOSPC901625-1GEA 435 EUR
APOSPC901625-1G
3,3,3-Trifluoropropanamide 95%
3,3,3-Trifluoropropanamide
Formel: C₃H₄F₃NO
Molecular Weight: 127,065 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11646085
CAS-Nummer: 460-75-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR