Methyl-2,3,4-trifluorbenzoat 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 2,3,4-trifluorobenzoate

PC901629-25G
APOSPC901629-25GEA 168 EUR
APOSPC901629-25G
Methyl-2,3,4-trifluorbenzoat 98%
Methyl-2,3,4-trifluorbenzoat
Formel: C₈H₅F₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06201866
CAS-Nummer: 773873-68-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR