4-Methyl-3-(trifluoromethyl)phenylboronic acid 95%

Lieferant: Apollo Scientific
Achtung

PC901644-1G PC901644-5G
APOSPC901644-1GEA 0 EUR
APOSPC901644-1G APOSPC901644-5G
4-Methyl-3-(trifluoromethyl)phenylboronic acid 95%
4-Methyl-3-(trifluoromethyl)phenylboronic acid
Formel: C₈H₈BF₃O₂
Lagertemperatur: Kühlschrank
MDL: MFCD11042431
CAS-Nummer: 947533-94-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR