2-Fluoro-4-isopropoxy-1-nitrobenzene 97%

Lieferant: Apollo Scientific

PC901656-1G
APOSPC901656-1GEA 270 EUR
APOSPC901656-1G
2-Fluoro-4-isopropoxy-1-nitrobenzene 97%
2-Fluoro-4-isopropoxy-1-nitrobenzene
Formel: C₉H₁₀FNO₃
Molecular Weight: 199,179 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115635
CAS-Nummer: 28987-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR