2,5-Difluoro-4-formylphenylboronic acid 95%
Lieferant: Apollo Scientific
Synonyme:
2,5-Difluoro-4-formylphenylphenylboronic acid, (2,5-Difluoro-4-formylphenyl)boronic acid
PC901710-1G
APOSPC901710-1GEA
217
EUR
APOSPC901710-1G
2,5-Difluoro-4-formylphenylboronic acid 95%
2,5-Difluoro-4-formylphenylboronic acid
Formel:
C₇H₅BF₂O₃ Molecular Weight: 185,921 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18383627 CAS-Nummer: 1228828-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...