2-(4-Fluorophenyl)-2-methylpropanenitrile 98%

Lieferant: Apollo Scientific

PC901813-1G
APOSPC901813-1GEA 429 EUR
APOSPC901813-1G
2-(4-Fluorophenyl)-2-methylpropanenitrile 98%
2-(4-Fluorophenyl)-2-methylpropanenitrile
Formel: C₁₀H₁₀FN
Molecular Weight: 163.192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11036570
CAS-Nummer: 93748-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR