4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole 97%

Lieferant: Apollo Scientific

PC901837-5G
APOSPC901837-5GEA 78.3 EUR
APOSPC901837-5G
4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole 97%
4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole
Formel: C₅H₄BrF₃N₂
Molecular Weight: 229.01 g/mol
Siedepunkt: 52…56 °C/0.5mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD04115415
CAS-Nummer: 497832-99-2

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR