2,2-Difluor-2-p-tolylessigsäure 98%
Lieferant: Apollo Scientific
PC901893-1G
APOSPC901893-1GEA
435
EUR
APOSPC901893-1G
2,2-Difluor-2-p-tolylessigsäure 98%
2,2-Difluor-2-p-tolylessigsäure
Formel:
C₉H₈F₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11007728 CAS-Nummer: 131323-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...