2-(4-Fluorphenyl)oxazol 97%
Lieferant: Apollo Scientific
PC901976-10G
APOSPC901976-10GEA
887
EUR
APOSPC901976-10G
2-(4-Fluorphenyl)oxazol 97%
2-(4-Fluorophenyl)oxazole
Formel:
C₉H₆FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234435 CAS-Nummer: 885268-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...