5-Bromo-1,2-difluoro-3-nitrobenzene 95%

Lieferant: Apollo Scientific

PC901988-5G
APOSPC901988-5GEA 75.4 EUR
APOSPC901988-5G
5-Bromo-1,2-difluoro-3-nitrobenzene 95%
5-Bromo-1,2-difluoro-3-nitrobenzene
Formel: C₆H₂BrF₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18311821
CAS-Nummer: 1261988-16-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR