1-(3-Fluorophenyl)cyclobutane-1-carboxylic acid 97%

Lieferant: Apollo Scientific

Synonyme: 1-(3-Fluorophenyl)cyclobutanecarboxylic acid

PC902033-1G
APOSPC902033-1GEA 30.6 EUR
APOSPC902033-1G
1-(3-Fluorophenyl)cyclobutane-1-carboxylic acid 97%
1-(3-Fluorophenyl)cyclobutane-1-carboxylic acid
Formel: C₁₁H₁₁FO₂
Molecular Weight: 194,202 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08444832
CAS-Nummer: 179411-84-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR