1-Chloro-2-nitro-4-(trifluoromethoxy)benzene

Lieferant: Apollo Scientific

PC902041-100G
APOSPC902041-100GEA 116 EUR
APOSPC902041-100G
1-Chloro-2-nitro-4-(trifluoromethoxy)benzene
1-Chloro-2-nitro-4-(trifluoromethoxy)benzene
Formel: C₇H₃ClF₃NO₃
Molecular Weight: 241.552 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194447
CAS-Nummer: 588-09-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR