2-Ethoxy-1-fluoro-4-nitrobenzene 97%
Lieferant: Apollo Scientific
PC902093-1G
APOSPC902093-1GEA
415
EUR
APOSPC902093-1G
2-Ethoxy-1-fluoro-4-nitrobenzene 97%
2-Ethoxy-1-fluoro-4-nitrobenzene
Formel:
C₈H₈FNO₃ Molecular Weight: 185,152 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20133797 CAS-Nummer: 1093656-34-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...