2-Fluoro-N-methyl-6-nitroaniline 98%
Lieferant: Apollo Scientific
PC902097-1G
APOSPC902097-1GEA
112
EUR
APOSPC902097-1G
2-Fluoro-N-methyl-6-nitroaniline 98%
2-Fluoro-N-methyl-6-nitroaniline
Formel:
C₇H₇FN₂O₂ Molecular Weight: 170.141 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23725947 CAS-Nummer: 182551-18-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...