2-(4-Fluorophenyl)-2-methylpropanoic acid 96%

Lieferant: Apollo Scientific

PC902103-1G
APOSPC902103-1GEA 444 EUR
APOSPC902103-1G
2-(4-Fluorophenyl)-2-methylpropanoic acid 96%
2-(4-Fluorophenyl)-2-methylpropanoic acid
Formel: C₁₀H₁₁FO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11036916
CAS-Nummer: 93748-19-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR