1-(4-Fluorophenyl)pyrazole 96%

Lieferant: Apollo Scientific

PC902184-1G
APOSPC902184-1GEA 121 EUR
APOSPC902184-1G
1-(4-Fluorophenyl)pyrazole 96%
1-(4-Fluorophenyl)pyrazole
Formel: C₉H₇FN₂
Molecular Weight: 162,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12192639
CAS-Nummer: 81329-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR