1-tert-Butyl-6-fluorbenzimidazol 98%

Lieferant: Apollo Scientific

PC902196-1G PC902196-5G PC902196-25G
APOSPC902196-1GEA 125 EUR
APOSPC902196-1G APOSPC902196-5G APOSPC902196-25G
1-tert-Butyl-6-fluorbenzimidazol 98%
1-tert-Butyl-6-fluorbenzimidazol
Formel: C₁₁H₁₃FN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12546476
CAS-Nummer: 1187385-71-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR