4-Fluor-2-mercaptobenzothiazol 98%
Lieferant: Apollo Scientific
Synonyme:
4-Fluorobenzo[d]thiazole-2(3H)-thione
PC902198-5G
APOSPC902198-5GEA
108
EUR
APOSPC902198-5G
4-Fluor-2-mercaptobenzothiazol 98%
4-Fluoro-2-mercaptobenzothiazole
Formel:
C₇H₄FNS₂ Molecular Weight: 185.25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06408877 CAS-Nummer: 154327-24-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...