6-Fluoro-1,3-benzoxazol-2-amine 98%

Lieferant: Apollo Scientific

Synonyme: 6-Fluorobenzo[d]oxazol-2-amine

PC902232-1G
APOSPC902232-1GEA 172 EUR
APOSPC902232-1G
6-Fluoro-1,3-benzoxazol-2-amine 98%
6-Fluoro-1,3-benzoxazol-2-amine
Formel: C₇H₅FN₂O
Molecular Weight: 152,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13188614
CAS-Nummer: 1199215-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR