1,5-Difluoro-2-methyl-4-nitrobenzene 96%

Lieferant: Apollo Scientific

Synonyme: 2,4-Difluoro-5-nitrotoluene

PC902246-1G PC902246-5G
APOSPC902246-1GEA 52.2 EUR
APOSPC902246-1G APOSPC902246-5G
1,5-Difluoro-2-methyl-4-nitrobenzene 96%
1,5-Difluoro-2-methyl-4-nitrobenzene
Formel: C₇H₅F₂NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16620288
CAS-Nummer: 179011-38-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR