1,5-Difluoro-2-methyl-4-nitrobenzene 96%
Lieferant: Apollo Scientific
Synonyme:
2,4-Difluoro-5-nitrotoluene
PC902246-1G
PC902246-5G
APOSPC902246-1GEA
52.2
EUR
APOSPC902246-1G
APOSPC902246-5G
1,5-Difluoro-2-methyl-4-nitrobenzene 96%
1,5-Difluoro-2-methyl-4-nitrobenzene
Formel:
C₇H₅F₂NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD16620288 CAS-Nummer: 179011-38-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...