4-(Trifluoromethyl)indole 97%
Lieferant: Apollo Scientific
PC902315-1G
PC902315-250MG
PC902315-5G
APOSPC902315-1GEA
0
EUR
APOSPC902315-1G
APOSPC902315-250MG
APOSPC902315-5G
4-(Trifluoromethyl)indole 97%
4-(Trifluoromethyl)indole
Formel:
C₉H₆F₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD09954766 CAS-Nummer: 128562-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...