2-(3-Fluorophenyl)benzonitrile 98%

Lieferant: Apollo Scientific

PC902361-1G PC902361-5G
APOSPC902361-1GEA 145 EUR
APOSPC902361-1G APOSPC902361-5G
2-(3-Fluorophenyl)benzonitrile 98%
2-(3-Fluorophenyl)benzonitrile
Formel: C₁₃H₈FN
Lagertemperatur: Raumtemperatur
MDL: MFCD05980284
CAS-Nummer: 1352318-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR