5-[3,5-Bis(trifluoromethyl)phenyl]-1,2,4-triazole-3-(2H)-thione 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC9025-1G
APOSPC9025-1GEA
0
EUR
APOSPC9025-1G
5-[3,5-Bis(trifluoromethyl)phenyl]-1,2,4-triazole-3-(2H)-thione 97%
5-[3,5-Bis(trifluoromethyl)phenyl]-1,2,4-triazole-3-(2H)-thione
Formel:
C₁₀H₅F₆N₃S Schmelzpunkt: 280 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052515 CAS-Nummer: 175276-77-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...