3,4-Difluoro-5-(trifluoromethyl)benzyl bromide 95%

Lieferant: Apollo Scientific

PC902686-1G
APOSPC902686-1GEA 0 EUR
APOSPC902686-1G
3,4-Difluoro-5-(trifluoromethyl)benzyl bromide 95%
3,4-Difluoro-5-(trifluoromethyl)benzyl bromide
Formel: C₈H₄BrF₅
Lagertemperatur: Raumtemperatur
MDL: MFCD00236263
CAS-Nummer: 239079-92-6
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR