3-Fluor-2-methoxy-6-methylpyridin 95%

Lieferant: Apollo Scientific

Synonyme: 3-Fluoro-2-methoxy-6-methylpyridine, 3-Fluoro-2-methoxy-6-picoline

PC902717-5G
APOSPC902717-5GEA 203 EUR
APOSPC902717-5G
3-Fluor-2-methoxy-6-methylpyridin 95%
3-Fluoro-2-methox-6-methylypyridine
Formel: C₇H₈FNO
Molecular Weight: 141.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03095083
CAS-Nummer: 375368-80-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR