1,2,3-Trifluoro-5-methylbenzene 95%

Lieferant: Apollo Scientific

PC902729-1G
APOSPC902729-1GEA 721 EUR
APOSPC902729-1G
1,2,3-Trifluoro-5-methylbenzene 95%
1,2,3-Trifluoro-5-methylbenzene
Formel: C₇H₅F₃
Molecular Weight: 146,11 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03425901
CAS-Nummer: 284463-96-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR