7-Fluoro-2-tetralone ≥98%

Lieferant: Apollo Scientific

Synonyme: 7-Fluoro-3,4-dihydro-1H-naphthalen-2-one

PC902738-1G PC902738-250MG
APOSPC902738-1GEA 543 EUR
APOSPC902738-1G APOSPC902738-250MG
7-Fluoro-2-tetralone ≥98%
7-Fluoro-2-tetralone
Formel: C₁₀H₉FO
Molecular Weight: 164.176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038615
CAS-Nummer: 29419-15-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR