3-Fluor-6-methyl-2-nitroanilin 96%
Lieferant: Apollo Scientific
|
Achtung
|
PC902766-1G
APOSPC902766-1GEA
364
EUR
APOSPC902766-1G
3-Fluor-6-methyl-2-nitroanilin 96%
3-Fluor-6-methyl-2-nitroanilin
Formel:
C₇H₇FN₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD22383663 CAS-Nummer: 1393442-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...