4-Fluoromethyl-pyridine 95%

Lieferant: Apollo Scientific

PC902794-1G
APOSPC902794-1GEA 487 EUR
APOSPC902794-1G
4-Fluoromethyl-pyridine 95%
4-Fluoromethyl-pyridine
Formel: C₆H₆FN
Molecular Weight: 111.117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08706067
CAS-Nummer: 82878-59-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR