N-(Chloroacetyl)-3-(trifluoromethyl)aniline

Lieferant: Apollo Scientific
Achtung

PC9028-25G PC9028-100G
APOSPC9028-25GEA 0 EUR
APOSPC9028-25G APOSPC9028-100G
N-(Chloroacetyl)-3-(trifluoromethyl)aniline
N-(Chloroacetyl)-3-(trifluoromethyl)aniline
Formel: C₉H₇ClF₃NO
Lagertemperatur: Raumtemperatur
CAS-Nummer: 351-38-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR