5-Chloro-2-fluoro-3-hydroxypyridine 95%

Lieferant: Apollo Scientific

PC902807-1G PC902807-250MG
APOSPC902807-1GEA 502 EUR
APOSPC902807-1G APOSPC902807-250MG
5-Chloro-2-fluoro-3-hydroxypyridine 95%
5-Chloro-2-fluoro-3-hydroxypyridine
Formel: C₅H₃ClFNO
Molecular Weight: 147.535 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09835255
CAS-Nummer: 1003711-65-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR