4,5-Difluor-2-methylbenzonitril 95%
Lieferant: Apollo Scientific
PC902816-5G
APOSPC902816-5GEA
98.6
EUR
APOSPC902816-5G
4,5-Difluor-2-methylbenzonitril 95%
4,5-Difluor-2-methylbenzonitril
Formel:
C₈H₅F₂N Molecular Weight: 153.1300 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09996935 CAS-Nummer: 1003708-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...